Learn More
MSC Steiner Welding Gloves, Split Cowhide ThermoCore, 14 in, 1 Pair.
Catalog No. 5000685451
Encompass
These general-duty stick welding gloves are designed for consistent quality and reliability. They are crafted from leather and synthetic leather, featuring a 14-inch length suitable for various welding applications.
  • Select shoulder split cowhide leather
  • ThermoCore foam insulated back with full cotton lining for comfort and heat protection
  • Palm reinforcement for added protection in high wear areas
  • Welted fingers and seams protect stitching
  • Wing thumb for natural feel and versatile movement
  • Durable lock stitching with flame resistant Kevlar thread for added strength
  • Gauntlet cuff style
  • Protection against abrasion, heat, and flame
Catalog No. 50-006-85451 Supplier MSC Supplier No. 42590729
These general-duty stick welding gloves are designed for consistent quality and reliability. They are crafted from leather and synthetic leather, featuring a 14-inch length suitable for various welding applications.
  • Select shoulder split cowhide leather
  • ThermoCore foam insulated back with full cotton lining for comfort and heat protection
  • Palm reinforcement for added protection in high wear areas
  • Welted fingers and seams protect stitching
  • Wing thumb for natural feel and versatile movement
  • Durable lock stitching with flame resistant Kevlar thread for added strength
  • Gauntlet cuff style
  • Protection against abrasion, heat, and flame