Learn More
MSC Steiner Welding Jacket, CF Carbonized Fiber, 2X-Large, 1 ea
Catalog No. 5000793699
Encompass
The CF Woven Carbonized Fiber Welding Jacket provides superior protection for welding applications. Made from carbonized fibers blended with para-aramid, this jacket is inherently flame resistant, tough, lightweight, breathable, and comfortable.
  • Features 10 oz woven carbonized fiber fabric for flame resistance
  • Relaxed fit with elongated front for comfort
  • Adjustable snap cuffs
  • Large inside pocket and sleeve pockets for tools
  • Stand-up/roll-down collar protects the neck
  • Black anodized non-reflective snaps
  • Sewn with flame resistant Nomex® thread
  • Provides 30 inch length for extended coverage
  • Complies with ASTM D6413-1999
Catalog No. 50-007-93699 Supplier MSC Supplier No. 67075556
The CF Woven Carbonized Fiber Welding Jacket provides superior protection for welding applications. Made from carbonized fibers blended with para-aramid, this jacket is inherently flame resistant, tough, lightweight, breathable, and comfortable.
  • Features 10 oz woven carbonized fiber fabric for flame resistance
  • Relaxed fit with elongated front for comfort
  • Adjustable snap cuffs
  • Large inside pocket and sleeve pockets for tools
  • Stand-up/roll-down collar protects the neck
  • Black anodized non-reflective snaps
  • Sewn with flame resistant Nomex® thread
  • Provides 30 inch length for extended coverage
  • Complies with ASTM D6413-1999