Learn More
MSC Steiner Welding Jacket, Cowhide, 30 In Length, Large, 1 Ea
Catalog No. 5000686236
Encompass
This welding jacket features a unique design with underarm ventilation and an open seam upper back, promoting increased air circulation for cool comfort in warm environments. It is constructed from premium side split brown cowhide and has a 30 in length.
  • Unique design with underarm ventilation and open seam upper back for increased air circulation
  • Made from premium side split brown cowhide
  • Satin-lined shoulders and upper arms for easy on and off
  • Stand-up/roll-down leather collar with secure snaps for neck protection
  • Inside pocket for spatter protection
  • Rivet reinforced seams and stress points
  • Lock stitched with flame resistant aramid fiber thread
Catalog No. 50-006-86236 Supplier MSC Supplier No. 63854798
This welding jacket features a unique design with underarm ventilation and an open seam upper back, promoting increased air circulation for cool comfort in warm environments. It is constructed from premium side split brown cowhide and has a 30 in length.
  • Unique design with underarm ventilation and open seam upper back for increased air circulation
  • Made from premium side split brown cowhide
  • Satin-lined shoulders and upper arms for easy on and off
  • Stand-up/roll-down leather collar with secure snaps for neck protection
  • Inside pocket for spatter protection
  • Rivet reinforced seams and stress points
  • Lock stitched with flame resistant aramid fiber thread