Learn More
MSC Steiner Welding Jacket, Large, Cotton, Flame-Resistant, 1 Each
Catalog No. 5000782907
Encompass
This large-sized cotton jacket is designed for welding applications, offering flame resistance and durability. It features a snaps closure and two pockets, providing practical functionality for industrial use.
  • Flame-resistant and flame-retardant
  • Anti-static properties
  • Resistant to rot and mildew
  • Snaps closure
  • Two pockets
  • Material weight: 9.00 oz
Catalog No. 50-007-82907 Supplier MSC Supplier No. 99019598
This large-sized cotton jacket is designed for welding applications, offering flame resistance and durability. It features a snaps closure and two pockets, providing practical functionality for industrial use.
  • Flame-resistant and flame-retardant
  • Anti-static properties
  • Resistant to rot and mildew
  • Snaps closure
  • Two pockets
  • Material weight: 9.00 oz