Learn More
Med Vet International Steinman Pin 1/8" x 9" w/ Trocar-Screw, J0251E, Each
Catalog No. 5000582082
Encompass_Preferred
Highest quality type 316L stainless steel IM pins. Available in the points and diameters listed below. When ordering, please match diameter with point style number. Made in U.S.A.
Catalog No. 50-005-82082 Supplier Med Vet International Supplier No. JOR251E
Highest quality type 316L stainless steel IM pins. Available in the points and diameters listed below. When ordering, please match diameter with point style number. Made in U.S.A.