Learn More
Sklar Instruments Steinmann Pin, Sklar, 4.5mm x 9 in. Steinmann Pin Trocar Point, EA/1.
SDP
Catalog No. 501438345 Shop All Sklar Instruments Products
Encompass_Preferred
Steinmann Pin, Sklar, 4.5mm x 9 in. Steinmann Pin Trocar Point, Sklar's Steinmann Pin is a thin metal rod commonly used in orthopedic procedures for holding large bone fractures together. These pins function similarly to K-Wire, but are larger in diameter. This product has a trocar point, 4.5mm diameter, and is 9 inches in length.
This item is not returnable. View return policy
Catalog No. 50-143-8345 Supplier Sklar Instruments Supplier No. 401596
Steinmann Pin, Sklar, 4.5mm x 9 in. Steinmann Pin Trocar Point, Sklar's Steinmann Pin is a thin metal rod commonly used in orthopedic procedures for holding large bone fractures together. These pins function similarly to K-Wire, but are larger in diameter. This product has a trocar point, 4.5mm diameter, and is 9 inches in length.