Learn More
Takara Bio Stellar™ Competent Cells (dam-/dcm-), 10 transformations
SDP
Catalog No. NC0609927
Encompass
Stellar Competent Cells (dam-/dcm-) are an E. coli HST04 strain that lacks the genetic factors (dam and dcm) necessary for the methylation of DNA. Plasmids prepared using this product can be cut by restriction enzymes which are normally blocked by dam or dcm methylation. A dam/recA double mutation is lethal, so the parent strain is recA+. Therefore, it follows that transformation of extracellular DNA with repeat sequences can result in recombination by recA. This product is not a suitable cloning host. For transformation, it is highly recommended to use this product for constructing plasmids as mentioned above. A pUC19 vector is provided as well as SOC Medium. 10 transformations
Catalog No. NC0609927 Supplier Takara Bio Supplier No. 636764
Stellar Competent Cells (dam-/dcm-) are an E. coli HST04 strain that lacks the genetic factors (dam and dcm) necessary for the methylation of DNA. Plasmids prepared using this product can be cut by restriction enzymes which are normally blocked by dam or dcm methylation. A dam/recA double mutation is lethal, so the parent strain is recA+. Therefore, it follows that transformation of extracellular DNA with repeat sequences can result in recombination by recA. This product is not a suitable cloning host. For transformation, it is highly recommended to use this product for constructing plasmids as mentioned above. A pUC19 vector is provided as well as SOC Medium. 10 transformations