Learn More
Takara Bio Stellar™ Competent Cells 50 x 100 ul
SDP
Catalog No. NC0197884
Encompass
Stellar Competent Cells are an E. coli HST08 strain that provides high transformation efficiency. These cells can be used in a wide variety of applications-from preparation of cDNA and genomic libraries, to construction of longer-length genomic libraries, to subcloning, and even methylated DNA cloning. Stellar Competent Cells lack the gene cluster for cutting foreign methylated DNA (mrr-hsdRMS-mcrBC and mcrA), and are therefore useful for cloning methylated DNA. The cells can also be used for blue/white screening (i.e., α-complementation) when transformed with vectors containing the lacZα gene. This package includes 50 tubes of competent cells (100 μl/tube), SOC Medium and a pUC19 vector. 50 x 100 ul
Catalog No. NC0197884 Supplier Takara Bio Supplier No. 636766
Stellar Competent Cells are an E. coli HST08 strain that provides high transformation efficiency. These cells can be used in a wide variety of applications-from preparation of cDNA and genomic libraries, to construction of longer-length genomic libraries, to subcloning, and even methylated DNA cloning. Stellar Competent Cells lack the gene cluster for cutting foreign methylated DNA (mrr-hsdRMS-mcrBC and mcrA), and are therefore useful for cloning methylated DNA. The cells can also be used for blue/white screening (i.e., α-complementation) when transformed with vectors containing the lacZα gene. This package includes 50 tubes of competent cells (100 μl/tube), SOC Medium and a pUC19 vector. 50 x 100 ul