Learn More
Sciencell Research Laboratories STELLATE CELL MEDIUM
Catalog No. NC9684692
Encompass
Stellate Cell medium is a complete medium designed for optimal growth of normal human stellate cells in vitro. It is shipped as a kit with stellate cell growth supplement and Pen/Strep solution.
Catalog No. NC9684692 Supplier Sciencell Research Laboratories Supplier No. 5301
Stellate Cell medium is a complete medium designed for optimal growth of normal human stellate cells in vitro. It is shipped as a kit with stellate cell growth supplement and Pen/Strep solution.