Learn More
Lonza Walkersville Stellate Cells Human Cryo 200k
Catalog No. 5000496248
Encompass
Stellate cells are a resident cell type of the liver primarily functioning to store retinoids In response to liver damage stellate cells rapidly lose the stored retinoids and differentiate into a proliferating fibroblast-like cell that begins depositing collagen matrix This activity causes buildup of collagen in the liver eventually leading to cirrhosis Stellate cells can be isolated from liver tissue enriched and placed into cell culture When placed into culture stellate cells begin to immediately differentiate into the activated state characterized by high expression of alpha-smooth muscle actin a-SMA Human Stellate Cells are stellate cells enriched from human liver isolations and grown to 80% confluency and then cryopreserved Human Stellate Growth Medium (Catalog MST-500BK) is available to support optimal culture health and growth characteristics of stellate cells Characterization performed Post-thaw plating to observe characteristic stellate morphology
Catalog No. 50-004-96248 Supplier Lonza Walkersville Supplier No. HUCLS200K
Stellate cells are a resident cell type of the liver primarily functioning to store retinoids In response to liver damage stellate cells rapidly lose the stored retinoids and differentiate into a proliferating fibroblast-like cell that begins depositing collagen matrix This activity causes buildup of collagen in the liver eventually leading to cirrhosis Stellate cells can be isolated from liver tissue enriched and placed into cell culture When placed into culture stellate cells begin to immediately differentiate into the activated state characterized by high expression of alpha-smooth muscle actin a-SMA Human Stellate Cells are stellate cells enriched from human liver isolations and grown to 80% confluency and then cryopreserved Human Stellate Growth Medium (Catalog MST-500BK) is available to support optimal culture health and growth characteristics of stellate cells Characterization performed Post-thaw plating to observe characteristic stellate morphology