Learn More
Lonza Walkersville Stellate cells human Passage 1
Catalog No. 5000496273
Encompass
Stellate cells are a resident cell type of the liver primarily functioning to store retinoids In response to liver damage stellate cells rapidly lose the stored retinoids and differentiate into a proliferating fibroblast-like cell that begins depositing collagen matrix This activity causes buildup of collagen in the liver eventually leading to cirrhosis Stellate cells can be isolated from liver tissue enriched and placed into cell culture When placed into culture stellate cells begin to immediately differentiate into the activated state Human Cryopreserved Stellate Cells 100K cells/vial are stellate cells enriched from human liver isolations and grown to 80% confluency and then cryopreserved Human Stellate Growth Medium (Catalog MCST250) is available to support optimal culture health and growth characteristics of stellate cells Characterization performed Post-thaw plating to observe characteristic stellate morphology
Catalog No. 50-004-96273 Supplier Lonza Walkersville Supplier No. HUCLS
Stellate cells are a resident cell type of the liver primarily functioning to store retinoids In response to liver damage stellate cells rapidly lose the stored retinoids and differentiate into a proliferating fibroblast-like cell that begins depositing collagen matrix This activity causes buildup of collagen in the liver eventually leading to cirrhosis Stellate cells can be isolated from liver tissue enriched and placed into cell culture When placed into culture stellate cells begin to immediately differentiate into the activated state Human Cryopreserved Stellate Cells 100K cells/vial are stellate cells enriched from human liver isolations and grown to 80% confluency and then cryopreserved Human Stellate Growth Medium (Catalog MCST250) is available to support optimal culture health and growth characteristics of stellate cells Characterization performed Post-thaw plating to observe characteristic stellate morphology