Learn More
Alpco Ltd Chemiluminescence Human Insulin ELISA
SDP
Supplier:  Alpco Ltd 80INSHUCH01
Encompass_Preferred
The Chemi Human Insulin ELISA measures the concentration of insulin in human serum, heparin plasma, and tissue culture supernatants.
  • Broad dynamic range of 5–30,000 pg/mL
  • Superior sensitivity compared other products on the market. 
  • LOD: <2.0 pg/mL
  • LLOQ: 20 pg/mL
FDA Class I Device. For In Vitro Diagnostic Use.
Catalog No. NC1446093