Learn More
ATCC Stem Cell Dissociation Reagent
Catalog No. 502384009
Encompass
Stem Cell Dissociation Reagent is a neutral protease isolated from Bacillus polymyxa and has applications in stem cell research. The reagent promotes safe and efficient detachment of human embryonic stem cells (hESC) and human induced pluripotent stem cells (hiPSC) during subcultivation in cell culture.
Catalog No. 50-238-4009 Supplier ATCC Supplier No. ACS3010
Stem Cell Dissociation Reagent is a neutral protease isolated from Bacillus polymyxa and has applications in stem cell research. The reagent promotes safe and efficient detachment of human embryonic stem cells (hESC) and human induced pluripotent stem cells (hiPSC) during subcultivation in cell culture.