Learn More
Cusabio Technology LLC Stem Cell Factor Receptor SCFR
Catalog No. 5000051787
Encompass_Preferred
This ELISA kit is designed for the quantitative detection of Human Stem Cell Factor Receptor (SCFR) in serum plasma and tissue homogenates Utilizing a sandwich assay format it provides a sensitive detection range of 1 56 ng/mL to 100 ng/mL with a lower limit of sensitivity at 0 39 ng/mL The assay requires a sample volume of 50-100 L and delivers results within 1-5 hours through absorbance measurement at 450 nm Ideal for research and clinical applications this kit ensures reliable measurement of SCFR concentrations supporting studies in cellular signaling hematopoietic regulation and disease biomarker analysis
Catalog No. 50-000-51787 Supplier Cusabio Technology LLC Supplier No. CSBE04721H
This ELISA kit is designed for the quantitative detection of Human Stem Cell Factor Receptor (SCFR) in serum plasma and tissue homogenates Utilizing a sandwich assay format it provides a sensitive detection range of 1 56 ng/mL to 100 ng/mL with a lower limit of sensitivity at 0 39 ng/mL The assay requires a sample volume of 50-100 L and delivers results within 1-5 hours through absorbance measurement at 450 nm Ideal for research and clinical applications this kit ensures reliable measurement of SCFR concentrations supporting studies in cellular signaling hematopoietic regulation and disease biomarker analysis