Learn More
ATCC Stem Cell Freezing Media
Catalog No. 502384623
Encompass
Stem Cell Freezing Media can be used in stem cell research, cell growth and viability, and cryopreservation of various types of human and non-human animal stem cells. This product has been qualified for use with primary-derived organoids, embryonic stem cells (hESC), induced pluripotent stem cells (hiPSC), and iPSCs-derived cells. Stem Cell Freezing Media can also be used with other continuous and cancer cell lines.
Catalog No. 50-238-4623 Supplier ATCC Supplier No. ACS3020
Stem Cell Freezing Media can be used in stem cell research, cell growth and viability, and cryopreservation of various types of human and non-human animal stem cells. This product has been qualified for use with primary-derived organoids, embryonic stem cells (hESC), induced pluripotent stem cells (hiPSC), and iPSCs-derived cells. Stem Cell Freezing Media can also be used with other continuous and cancer cell lines.