Learn More
Enzo Life Sciences SCREEN-WELL Stem Cell Library (100µl/well)
SDP
Catalog No. 502013865
Encompass_Preferred
The SCREEN-WELL Stem Cell Library is a collection of 130 compounds that can be used for stem cell signaling research and drug screening. The compounds are predominantly dissolved in DMSO at 10 mM and aliquoted into 96 deep-well plates at 100 µL per well. Long Term Storage: -80°C. Applications: HTS .
Catalog No. 50-201-3865 Supplier Enzo Life Sciences Supplier No. ENZLIB1030100
Add to Cart
Add to Cart
Request a Quote
The SCREEN-WELL Stem Cell Library is a collection of 130 compounds that can be used for stem cell signaling research and drug screening. The compounds are predominantly dissolved in DMSO at 10 mM and aliquoted into 96 deep-well plates at 100 µL per well. Long Term Storage: -80°C. Applications: HTS .