Learn More
Iwai North America STEM-CELLBANKER DMSO Free GMP grade: Ready to use serum and DMSO free GMP grade cryopreservation media. Size: 20mL
Catalog No. NC2733008
Encompass_Preferred
STEM-CELLBANKER DMSO Free GMP grade is a part of the renowned CELLBANKER series, cited in over 1400 publications. As part of Japan's most trusted line of cryopreservation media, is DMSO free, serum free, is ready to use, and requires no liquid nitrogen for cell preservation. Trust STEM-CELLBANKER DMSO free GMP grade for safe, hassle-free cell storage. **Registered with FDA DMF**
Catalog No. NC2733008 Supplier Iwai North America Supplier No. 13925
STEM-CELLBANKER DMSO Free GMP grade is a part of the renowned CELLBANKER series, cited in over 1400 publications. As part of Japan's most trusted line of cryopreservation media, is DMSO free, serum free, is ready to use, and requires no liquid nitrogen for cell preservation. Trust STEM-CELLBANKER DMSO free GMP grade for safe, hassle-free cell storage. **Registered with FDA DMF**