Learn More
Amsbio LLC STEM-CELLBANKER DMSO FREE - GMP Grade  (AOF Cryopreservation Media) (100ml)(formerly 11890F)
Catalog No. 502566877
Encompass
STEM-CELLBANKER DMSO free GMP Grade is a ready-to-use, chemically defined, animal-free stem cell cryopreservation solution manufactured under GMP conditions. It is optimized for embryonic stem (ES) and induced pluripotent stem (iPS) cell storage, as well as being a suitable solution for the cryopreservation of other fragile cell types. Containing only EU, US Pharmacopoeia graded ingredients, STEM-CELLBANKER DMSO free is the optimal choice for storage of cells developed for cell therapy applications. It is also available as a DMSO free formulation. MAIN FEATURES: Chemically defined, animal-free, GMP grade cryopreservation medium | Enables long term cell storage >10 years at -80˚C or -196˚C | Maintains cell pluripotency, normal karyotype, and proliferation ability post-thaw | FDA Master File registered | Suitable for cell, organoid, and tissue cryopreservation | Consistent high cell viability | No programmed freezer or liquid nitrogen required | DMSO free.
Catalog No. 50-256-6877 Supplier Amsbio LLC Supplier No. 13926
STEM-CELLBANKER DMSO free GMP Grade is a ready-to-use, chemically defined, animal-free stem cell cryopreservation solution manufactured under GMP conditions. It is optimized for embryonic stem (ES) and induced pluripotent stem (iPS) cell storage, as well as being a suitable solution for the cryopreservation of other fragile cell types. Containing only EU, US Pharmacopoeia graded ingredients, STEM-CELLBANKER DMSO free is the optimal choice for storage of cells developed for cell therapy applications. It is also available as a DMSO free formulation. MAIN FEATURES: Chemically defined, animal-free, GMP grade cryopreservation medium | Enables long term cell storage >10 years at -80˚C or -196˚C | Maintains cell pluripotency, normal karyotype, and proliferation ability post-thaw | FDA Master File registered | Suitable for cell, organoid, and tissue cryopreservation | Consistent high cell viability | No programmed freezer or liquid nitrogen required | DMSO free.