Learn More
Amsbio LLC STEM-CELLBANKER EX  - GMP Grade (AOF Cryopreservation Media for Intravenous Administration) (100ml)(formerly 11890EX)
Catalog No. 502566881
Encompass
STEM-CELLBANKER EX – GMP Grade is a chemically defined, xeno-free, cryopreservation media that only includes components registered as "Inactive Ingredients" by the FDA for intravenous administration. It is optimized for hematopoietic cells, STEM-CELLBANKER EX is suitable for the cryopreservation all types for derived therapeutic cells. MAIN FEATURES: All components suitable for intravenous application | Chemically defined, human and animal product free, GMP grade | Enables long term cell storage >10 years at -80˚C or -196˚C | FDA - CBER Master File registered | Suitable for cell, organoid, and tissue cryopreservation | Consistent high cell viability & recovery | No programmed freezer or liquid nitrogen required.
Catalog No. 50-256-6881 Supplier Amsbio LLC Supplier No. 11936
STEM-CELLBANKER EX – GMP Grade is a chemically defined, xeno-free, cryopreservation media that only includes components registered as "Inactive Ingredients" by the FDA for intravenous administration. It is optimized for hematopoietic cells, STEM-CELLBANKER EX is suitable for the cryopreservation all types for derived therapeutic cells. MAIN FEATURES: All components suitable for intravenous application | Chemically defined, human and animal product free, GMP grade | Enables long term cell storage >10 years at -80˚C or -196˚C | FDA - CBER Master File registered | Suitable for cell, organoid, and tissue cryopreservation | Consistent high cell viability & recovery | No programmed freezer or liquid nitrogen required.