Learn More
Iwai North America STEM-CELLBANKER EX GMP grade: Ready to use serum free GMP grade cryopreservation media. Size: 100mL
Catalog No. NC2733011
Encompass_Preferred
STEM-CELLBANKER EX GMP grade is a part of the renowned CELLBANKER series, cited in over 1400 publications. As part of Japan's most trusted line of cryopreservation media, this version is optimized for hHSC and human dispose-derived stromal/stem cells. It is serum free, is ready to use, and requires no liquid nitrogen for cell preservation. Trust STEM-CELLBANKER GMP grade for safe, hassle-free cell storage. **Registered with FDA DMF**
Catalog No. NC2733011 Supplier Iwai North America Supplier No. 11936
STEM-CELLBANKER EX GMP grade is a part of the renowned CELLBANKER series, cited in over 1400 publications. As part of Japan's most trusted line of cryopreservation media, this version is optimized for hHSC and human dispose-derived stromal/stem cells. It is serum free, is ready to use, and requires no liquid nitrogen for cell preservation. Trust STEM-CELLBANKER GMP grade for safe, hassle-free cell storage. **Registered with FDA DMF**