Learn More
Iwai North America STEM-CELLBANKER GMP grade: Ready to use serum free and GMP grade cryopreservation media. Size: 100mL
Catalog No. NC2732998
Encompass_Preferred
STEM-CELLBANKER GMP gradeis a part of the renowned CELLBANKER series, cited in over 1400 publications. As part of Japan's most trusted line of cryopreservation media, this GMP grade version is optimized for stem cells and your most delicate cells. It is serum free, is ready to use, and requires no liquid nitrogen for cell preservation. Trust STEM-CELLBANKER GMP grade for safe, hassle-free cell storage. **Registered with FDA DMF**
Catalog No. NC2732998 Supplier Iwai North America Supplier No. 11924
STEM-CELLBANKER GMP gradeis a part of the renowned CELLBANKER series, cited in over 1400 publications. As part of Japan's most trusted line of cryopreservation media, this GMP grade version is optimized for stem cells and your most delicate cells. It is serum free, is ready to use, and requires no liquid nitrogen for cell preservation. Trust STEM-CELLBANKER GMP grade for safe, hassle-free cell storage. **Registered with FDA DMF**