Learn More
K'NEX™ STEM Explorations: Levers & Pulleys Building Set
SCI_ED
Catalog No. S81356
Change view
Click to view available options
Educational Toy
Catalog No.
S81356 Educational Toy
1 options
Catalog No. S81356 Supplier K'NEX™ Supplier No. 79319
Add to Cart
Add to Cart
Levers and Pulleys Building Set allows parents and teachers to guide children in learning about lever classes, pulley systems, resistance, effort, and mechanical advantage.
  • 139 parts to build unique, fully-functioning replicas of real-world models: a balance, a wheelbarrow, and a sailboat
  • Includes 3 after-build experiments that teach your child about STEM concepts like lever classes, pulley systems, effort and more
  • Accommodates 1-2 students, working as a team
  • Ideal solution for home-school families
  • Easy-to-follow, step-by-step building instructions included
  • Building with K'NEX™ puts children on a path towards a fundamental understanding of STEAM/STEM subjects
  • K'NEX™ Education sets come with lesson plans and experiment guides written by educators that feature hands-on, inquiry-based projects
  • All teacher guides and experiment guides are aligned to national standards, including ITEEA, NSES, NCTM, NGSS and Common Core
  • K'NEX™ Education sets let students build replicas of real-world machines to gain an understanding of the principles that make them work
Specifications
Includes 139 parts and 3 after-build experiments
Product Type Education Toy