Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

K'NEX Education™ STEM Explorations: Roller Coaster Building Set
SCI_ED

Catalog No. S14016
Change view
Click to view available options
Grade:
5 to 9
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S14016 5 to 9
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S14016 Supplier K'NEX Education™ Supplier No. 77078

missing translation for 'validMainframeMsg'

Requires

2 AA batteries

Students explore STEM concepts while building a working roller coaster.

Using the materials included in this set, middle-school aged children will be engaged and energized as they further their knowledge and understanding of the science, technology, engineering and math concepts associated with a real-life amusement park ride.

 

Once built, a downloadable booklet guides students through 3 hands-on, inquiry-based experiments on each of the models. Students will learn about the relationship between time, distance and speed plus much more.

Set includes:

  • More than 500 parts plus a battery-powered motor for a single child or team of 2-3 children to build a working Roller Coaster
  • Two additional models: a Ramp and a Half-Pipe (each model can be built one at a time)

The Experiment Guide is aligned to national STEM standards and is appropriate for grade levels 5 to 9.

Specifications

Certifications/Compliance Experiment Guide is aligned to national STEM standards
Grade 5 to 9
Age 8
For Use With (Application) Teaching STEM concepts
Includes 500 parts, battery-powered motor, 2 additional models: ramp and half-piple
Requires 2 “AA” (LR6) Batteries not included
Class Size Team of 2 to 3 Students
Product Type Rollar coaster building set
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.