Learn More
Beckman Coulter Beckman Coulter AQUIOS STEM Kit Reagents - CD34+ Stem Cell Enumeration, 50 Tests
Catalog No. NC3538632
Encompass
The AQUIOS STEM Kit is an IVD reagent kit for identification and enumeration of CD34+ hematopoietic stem and progenitor cells by flow cytometry. The kit provides automated-compatible reagents, barcoded traceability, and absolute counting beads to support ISHAGE-compliant analysis.
  • Intended for identification and enumeration of CD34+ hematopoietic stem and progenitor cells.
  • Configured for 50 tests per kit.
  • Includes room-temperature ready lysing reagent and absolute counting beads.
  • Barcoded reagents and assay sheet for full traceability of reagents and specimens.
  • Supports automated sample preparation and analysis with ISHAGE-guided gating algorithms.
  • Provides two-level process controls that encompass clinical decision levels.
  • Enables duplicate runs, negative-control options, and single-test workflow flexibility.
  • Designed for room-temperature processing of samples and reagents.
Catalog No. NC3538632 Supplier Beckman Coulter Supplier No. IM3630US
The AQUIOS STEM Kit is an IVD reagent kit for identification and enumeration of CD34+ hematopoietic stem and progenitor cells by flow cytometry. The kit provides automated-compatible reagents, barcoded traceability, and absolute counting beads to support ISHAGE-compliant analysis.
  • Intended for identification and enumeration of CD34+ hematopoietic stem and progenitor cells.
  • Configured for 50 tests per kit.
  • Includes room-temperature ready lysing reagent and absolute counting beads.
  • Barcoded reagents and assay sheet for full traceability of reagents and specimens.
  • Supports automated sample preparation and analysis with ISHAGE-guided gating algorithms.
  • Provides two-level process controls that encompass clinical decision levels.
  • Enables duplicate runs, negative-control options, and single-test workflow flexibility.
  • Designed for room-temperature processing of samples and reagents.