Learn More
Cusabio Technology LLC stem-loop binding protein
Catalog No. 5000049824
Encompass_Preferred
This kit detects SLBP (Histone RNA hairpin-binding protein) in human serum plasma tissue homogenates and cell lysates using a sandwich ELISA method It offers a quantitative detection range of 15 6 pg/mL to 1000 pg/mL with a sensitivity of 3 9 pg/mL ensuring precise measurement of low-abundance samples The assay requires 50-100 L of sample per test and delivers results within 1-5 hours Optical density is measured at 450 nm for accurate quantification making it suitable for research applications requiring reliable assessment of SLBP levels in various biological matrices The standardized protocol and consistent performance enable efficient analysis of histone-related RNA-binding protein expression in experimental or diagnostic settings
Catalog No. 50-000-49824 Supplier Cusabio Technology LLC Supplier No. CSBEL021372HU
This kit detects SLBP (Histone RNA hairpin-binding protein) in human serum plasma tissue homogenates and cell lysates using a sandwich ELISA method It offers a quantitative detection range of 15 6 pg/mL to 1000 pg/mL with a sensitivity of 3 9 pg/mL ensuring precise measurement of low-abundance samples The assay requires 50-100 L of sample per test and delivers results within 1-5 hours Optical density is measured at 450 nm for accurate quantification making it suitable for research applications requiring reliable assessment of SLBP levels in various biological matrices The standardized protocol and consistent performance enable efficient analysis of histone-related RNA-binding protein expression in experimental or diagnostic settings