Learn More
Reprocell Stem RNA 3rd Gen Reprogramming Kit, Non-integrating, mRNA-based, 25,000 cells, Fibroblasts: 10-14 days, EPCs: 12-16 days (12-14), UPCs: 12-16 days (12-14), 4 transfections (Fibroblasts), 6-8 transfections (Urine/Blood)
Catalog No. NC2168572
Encompass
Stem RNA 3rd Gen Reprogramming Kit, Non-integrating, mRNA-based, 25,000 cells, Fibroblasts: 10-14 days, EPCs: 12-16 days (12-14), UPCs: 12-16 days (12-14), 4 transfections (Fibroblasts), 6-8 transfections (Urine/Blood)
This item is not returnable. View return policy
Catalog No. NC2168572 Supplier Reprocell Supplier No. 000076
Stem RNA 3rd Gen Reprogramming Kit, Non-integrating, mRNA-based, 25,000 cells, Fibroblasts: 10-14 days, EPCs: 12-16 days (12-14), UPCs: 12-16 days (12-14), 4 transfections (Fibroblasts), 6-8 transfections (Urine/Blood)