Learn More
STEMCELL Technologies StemSpan™ CD34+ Expansion Supplement (10X), Size: 10 mL
SDP
Catalog No. NC2039770
Encompass
StemSpan™ CD34+ Expansion Supplement (10X) contains a combination of recombinant human cytokines and other additives formulated to selectively promote the expansion of CD34+ cells isolated from human cord blood (CB) or bone marrow (BM) samples.StemSpan™ CD34+ Expansion Supplement typically promotes ~40-fold expansion of total nucleated cells in 7-day liquid cultures of CD34+ human cord blood (CB) cells. After one week, ~40% of the cultured cells express CD34, indicative of >10-fold expansion of input CD34+ CB cells. This expansion may be further increased with the addition of small molecules such as UM729. See data tab for more details. StemSpan™ CD34+ Expansion Supplement (10X) is intended for use in combination with any of the following StemSpan™ media:• StemSpan™ SFEM (Catalog #09600)• StemSpan™ SFEM II (Catalog #09605)• StemSpan™-XF (Catalog #100-0073)• StemSpan™-AOF (Catalog #100-0130)
This item is not returnable. View return policy
Catalog No. NC2039770 Supplier STEMCELL Technologies Supplier No. 02691
StemSpan™ CD34+ Expansion Supplement (10X) contains a combination of recombinant human cytokines and other additives formulated to selectively promote the expansion of CD34+ cells isolated from human cord blood (CB) or bone marrow (BM) samples.StemSpan™ CD34+ Expansion Supplement typically promotes ~40-fold expansion of total nucleated cells in 7-day liquid cultures of CD34+ human cord blood (CB) cells. After one week, ~40% of the cultured cells express CD34, indicative of >10-fold expansion of input CD34+ CB cells. This expansion may be further increased with the addition of small molecules such as UM729. See data tab for more details. StemSpan™ CD34+ Expansion Supplement (10X) is intended for use in combination with any of the following StemSpan™ media:• StemSpan™ SFEM (Catalog #09600)• StemSpan™ SFEM II (Catalog #09605)• StemSpan™-XF (Catalog #100-0073)• StemSpan™-AOF (Catalog #100-0130)