Learn More
Grainger New Age Industrial Swivel Stem Caster, Polyolefin, 5 in., Side Brake, 1 ea.
Catalog No. 502786471
Encompass_Preferred
This swivel stem caster is designed for seamless mobility and transport of heavy-duty equipment such as mobile shelving units. Engineered for durability, this caster features a 5 in. diameter polyolefin wheel, a zinc-plated frame, and a side brake for enhanced safety and stability during operation.
  • Features a 300 lb. load capacity
  • Swivel design with friction ring stem for easy maneuverability
  • Equipped with a side brake for secure positioning
  • Constructed with a non-marking polyolefin wheel
  • Designed for use in a temperature range of -30°F to 140°F
  • NSF listed for sanitation standards
Catalog No. 50-278-6471 Supplier Grainger Supplier No. 18K925
This swivel stem caster is designed for seamless mobility and transport of heavy-duty equipment such as mobile shelving units. Engineered for durability, this caster features a 5 in. diameter polyolefin wheel, a zinc-plated frame, and a side brake for enhanced safety and stability during operation.
  • Features a 300 lb. load capacity
  • Swivel design with friction ring stem for easy maneuverability
  • Equipped with a side brake for secure positioning
  • Constructed with a non-marking polyolefin wheel
  • Designed for use in a temperature range of -30°F to 140°F
  • NSF listed for sanitation standards