Learn More
Oz Biosciences STEM TRANSFECTION REG 1ML
SDP
Catalog No. NC3652768
Encompass
Stem Transfection Reagent is a powerful reagent allowing multipotent stem cells transfection with high efficiency and very low toxicity. Its specific composition based on the TEE-technology allows transfecting embryonic and multipotent stem cells in presence of serum maintaining their undifferentiated stage and their capacities to differentiate.
Catalog No. NC3652768 Supplier Oz Biosciences Supplier No. ST31000
Stem Transfection Reagent is a powerful reagent allowing multipotent stem cells transfection with high efficiency and very low toxicity. Its specific composition based on the TEE-technology allows transfecting embryonic and multipotent stem cells in presence of serum maintaining their undifferentiated stage and their capacities to differentiate.