Learn More
Takara Bio STEM121, 50 ug
SDP
Supplier:  Takara Bio Y40410
Encompass
STEM121 reacts with a human cytoplasmic protein expressed in brain, liver, pancreas, and CNS cells; the antibody does not cross-react with brain tissue or cell extracts from mouse, rat, or cynomolgous monkey.
Catalog No. NC1916110