Learn More
Stemculture LLC StemCultures LLC StemBeads GDNF
SDP
Supplier:  Stemculture LLC SBGD1
Encompass
StemBeads GDNF is a patented growth factor supplement that offers a novel way to culture cells with native Glial Derived Neurotrophic Factor more efficiently. GDNF is key for supporting neural progenitor cells (NPCs) and neuronal differentiation. StemBeads GDNF are microbeads composed of an FDA approved, biodegradable PLGA polymer that is loaded with native, non-mutant recombinant human GDNF and release the encapsulated protein at a constant rate. Under the microscope, StemBeads will appear as small dark spheres that do not harm the cells, and with time, will break down while releasing the encapsulated protein. Controlled delivery and stable levels overcome the 45-minute half-life of GDNF and improve cell cultures while saving researchers valuable time and resources. StemBeads GDNF has been tested in multiple neuronal base media, including neural progenitor expansion media (NPEM), with enhanced cellular profiles. StemBeads GDNF were used with and without StemBeads BDNF.
Catalog No. NC2641086