Learn More
MilliporeSigma™ STEMCCA Constitutive Polycistronic (OKSM) Lentivirus Reprogramming Kit
Catalog No. SCR530 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
SCR530 1 Kit
1 options
Catalog No. SCR530 Supplier MilliporeSigma™ Supplier No. SCR530
Add to Cart
Add to Cart
Contains high titer polycistronic lentivirus and Polybrene transfection reagent that have been validated for generation of mouse and human induced pluripotent stem cells. MilliporeSigma™; STEMCCA Constitutive Polycistronic (OKSM) Lentivirus Reprogramming Kit is comprised of the transcription factors Oct-4, Klf4, SOX-2, and c-Myc (OKSM), separated by the self-cleaving 2A peptide and IRES sequences.
  • Use of a single polycistronic lentiviral vector significantly improves reprogramming efficiencies and reduces the number of viral integrations
  • EF1α-STEMCCA (OKSM) Lentivirus: (Part number CS204279) Three (3) vials containing 15μL of high titer lentivirus
  • Polybrene Transfection Reagent: (Part number TR-1003-50UL) One (1) vial containing 50μL of 10mg/mL stock
TRUSTED_SUSTAINABILITY
Specifications
Product Type Lentivirus Reprogramming Kit
For Use With (Application) Stem Cell Culture
Quantity 1 Kit
Regulatory Status RUO
Content And Storage Lentivirus is stable for at least 6 months when stored at -80°C. After first thaw, place immediately on ice and store in working aliquots to avoid further freeze thaws. Avoid freeze thaws as this will result in a decrease in the virus titer. Polybrene transfection reagent is stable for at least 1 year when stored at -20°C.