Learn More
MilliporeSigma™ STEMCCA Constitutive Polycistronic (OKSM) Lentivirus Reprogramming Kit
Catalog No. SCR510 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
SCR510 1 Kit
1 options
Catalog No. SCR510 Supplier MilliporeSigma™ Supplier No. SCR510
Add to Cart
Add to Cart
Contains high titer polycistronic (OKSM) lentivirus and Polybrene transfection reagent that have been validated for the generation of mouse and human induced pluripotent stem cells from mouse embryonic fibroblasts (MEFs) and human foreskin fibroblasts (HFFs).
  • STEMCCA vector is comprised of transcription factors Oct-4, Klf4, SOX-2, and c-Myc (OKSM), separated by the self-cleaving 2A peptide and IRES sequences
  • Use of single polycistronic lentiviral vector significantly improves reprogramming efficiencies
  • Reduces number of viral integrations
Components:
  • EF1α-STEMCCA (OKSM) Lentivirus: (Part number CS204279) One (1) vial containing 15μL of high titer lentivirus
  • Polybrene Transfection Reagent: (Part number TR-1003-50UL) One (1) vial containing 50μL of 10mg/mL stock
TRUSTED_SUSTAINABILITY
Specifications
Product Type Lentivirus Reprogramming Kit
Quantity 1 Kit
Regulatory Status RUO
Content And Storage Stable for 3 months from date of receipt when stored at -20°C or -80°C. Upon first thaw, centrifuge the vial and gently mix the solution. Aliquot into smaller working volumes and freeze at -20°C or -80°C.