Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ STEMCCA Cre-Excisable Constitutive Polycistronic (OKSM) Lentivirus Reprogramming Kit

Catalog No. SCR531 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCR531 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCR531 Supplier MilliporeSigma™ Supplier No. SCR531
Add to Cart
Add to Cart

Contains high titer polycistronic lentivirus and Polybrene transfection reagent that have been validated for the generation of mouse and human induced pluripotent stem cells. MilliporeSigma™; STEMCCA Cre-Excisable Constitutive Polycistronic (OKSM) Lentivirus Reprogramming Kit comprises of the transcription factors Oct-4, Klf4, SOX-2, and c-Myc (OKSM) separated by the self-cleaving 2A peptide and IRES sequences.

  • Use of single polycistronic lentiviral vector significantly improves reprogramming efficiencies
  • Reduces the number of viral integrations
  • Components:
  • EF1α-STEMCCA-LoxP (OKSM) Lentivirus: (Part number CS204439) Three (3) vials containing 15μL of high titer lentivirus
  • Polybrene Transfection Reagent: (Part number TR-1003-50UL) One (1) vial containing 50μL of 10mg/mL stock
TRUSTED_SUSTAINABILITY

Specifications

Product Type Lentivirus Reprogramming Kit
For Use With (Application) Stem Cell Culture
Quantity 1 Kit
Regulatory Status RUO
Content And Storage Lentivirus is stable for at least 6 months when stored at -80°C. After first thaw, place immediately on ice and store in working aliquots to avoid further freeze thaws. Avoid freeze thaws as this will result in a decrease in the virus titer. Polybrene transfection reagent is stable for at least 1 year when stored at -20°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.