Learn More
MilliporeSigma™ STEMCCA™ Human Cre-Excisable Polycistronic (OKS/L-Myc) Lentivirus Reprogramming Kit
Catalog No. SCR548 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
SCR548 1 Kit
1 options
Catalog No. SCR548 Supplier MilliporeSigma™ Supplier No. SCR548
For stem cell culture applications
TRUSTED_SUSTAINABILITY
Specifications
Product Type Lentivirus Reprogramming Kit
For Use With (Application) Stem Cell Culture
Quantity 1 Kit
Content And Storage 1. One EF1a-hSTEMCCA-LoxP (OKS/L-Myc) Lentivirus: One (1) vial containing 15μL of high titer lentivirus2. Five Polybrene 10mg/mL: One (1) vial containing 50μL of 10 mg/mL stock of Polybrene transfection reagent3. Four PS48 Supplement (1000X): One (1) vial containing 400μL of the inhibitor in high quality DMSO4. Three Sodium Butyrate Supplement (1000X): One (1) vial containing 400μL of the inhibitor in water5. Two TGF-β RI Kinase Inhibitor IV Supplement (1000X): One (1) vial containing 400μL of the inhibitor in high quality DMSO. Lentivirus is stable for at least 6 months when stored at -80°C.