Learn More
STEMCELL Technologies STEMdiff™ Cardiomyocyte Freezing Medium, 50 mL
SDP
Catalog No. NC1539751
Encompass
STEMdiff™ Cardiomyocyte Freezing Medium can be used to cryopreserve cardiomyocytes derived from human pluripotent stem cells (hPSCs) and maintained in STEMdiff™ Cardiomyocyte Maintenance Medium (Catalog #05020). STEMdiff™ Cardiomyocyte Dissociation Kit (Catalog #05025) is required for dissociating cardiomyocytes prior to freezing. After thawing using STEMdiff™ Cardiomyocyte Support Medium (Catalog #05027), the cardiomyocytes can be used in various downstream applications and analyses. This medium supports hPSC-derived cardiomyocytes generated using STEMdiff™ Cardiomyocyte Differentiation Kit (Catalog #05010).
Catalog No. NC1539751 Supplier STEMCELL Technologies Supplier No. 05030
STEMdiff™ Cardiomyocyte Freezing Medium can be used to cryopreserve cardiomyocytes derived from human pluripotent stem cells (hPSCs) and maintained in STEMdiff™ Cardiomyocyte Maintenance Medium (Catalog #05020). STEMdiff™ Cardiomyocyte Dissociation Kit (Catalog #05025) is required for dissociating cardiomyocytes prior to freezing. After thawing using STEMdiff™ Cardiomyocyte Support Medium (Catalog #05027), the cardiomyocytes can be used in various downstream applications and analyses. This medium supports hPSC-derived cardiomyocytes generated using STEMdiff™ Cardiomyocyte Differentiation Kit (Catalog #05010).