Learn More
STEMCELL Technologies STEMdiff™ Cardiomyocyte Maintenance Kit, Size: 1 Kit
SDP
Catalog No. NC1663495
Encompass
STEMdiff™ Cardiomyocyte Maintenance Kit can be used for long-term maintenance of human pluripotent stem cell (PSC)-derived cardiomyocytes for one month or longer. The functional capacity of hPSC-derived cardiomyocytes is retained for downstream applications and analysis. This kit is compatible with cardiomyocytes generated using STEMdiff™ Ventricular Cardiomyocyte Differentiation Kit (Catalog #05010) or STEMdiff™ Atrial Cardiomyocyte Differentiation Kit (Catalog #100-0215).
Catalog No. NC1663495 Supplier STEMCELL Technologies Supplier No. 05020
STEMdiff™ Cardiomyocyte Maintenance Kit can be used for long-term maintenance of human pluripotent stem cell (PSC)-derived cardiomyocytes for one month or longer. The functional capacity of hPSC-derived cardiomyocytes is retained for downstream applications and analysis. This kit is compatible with cardiomyocytes generated using STEMdiff™ Ventricular Cardiomyocyte Differentiation Kit (Catalog #05010) or STEMdiff™ Atrial Cardiomyocyte Differentiation Kit (Catalog #100-0215).