Learn More
STEMCELL Technologies STEMdiff™ Cerebral Organoid Kit
SDP
Catalog No. NC1619403
Encompass
Generate self-organized, pluripotent stem cell (PSC)-derived neural organoids with a cellular composition and structural organization representative of the developing human brain. These defined, serum-free cell culture media and the simple, four-stage protocol are based on the formulation published by Lancaster et al. (Lancaster MA et al. Nature, 2013 and Lancaster MA et al. Science, 2014) to more reliably generate cerebral organoids. Beginning with an embryoid body formation step followed by expansion of neuroepithelia, organoids generated using STEMdiff™ Cerebral Organoid Kit feature cortical-like regions, including the ventricular zone (PAX6+/SOX2+/Ki-67+), outer subventricular zone (Ki-67+/p-Vimentin+), intermediate zone (TBR2+), and cortical plate (CTIP2+/MAP2+/TBR1+), which layer in similar orientations as those observed in vivo.
Catalog No. NC1619403 Supplier STEMCELL Technologies Supplier No. 08570
Generate self-organized, pluripotent stem cell (PSC)-derived neural organoids with a cellular composition and structural organization representative of the developing human brain. These defined, serum-free cell culture media and the simple, four-stage protocol are based on the formulation published by Lancaster et al. (Lancaster MA et al. Nature, 2013 and Lancaster MA et al. Science, 2014) to more reliably generate cerebral organoids. Beginning with an embryoid body formation step followed by expansion of neuroepithelia, organoids generated using STEMdiff™ Cerebral Organoid Kit feature cortical-like regions, including the ventricular zone (PAX6+/SOX2+/Ki-67+), outer subventricular zone (Ki-67+/p-Vimentin+), intermediate zone (TBR2+), and cortical plate (CTIP2+/MAP2+/TBR1+), which layer in similar orientations as those observed in vivo.