Learn More
STEMCELL Technologies STEMdiff™ Cerebral Organoid Maturation Kit, Size: 1 Kit
SDP
Catalog No. 501974446
Encompass
Culture pluripotent stem cell (PSC)-derived neural organoids for extended periods (> 40 days) with the STEMdiff™ Cerebral Organoid Maturation Kit.For your convenience, the maturation kit is fully compatible with the STEMdiff™ Cerebral Organoid Kit (/products/stemdiff-cerebral-organoid-kit.html), which can be used to generate neural organoids under defined and serum-free conditions.For more information on protocols for organoid culture with STEMdiff™ Cerebral Organoid Maturation Kit, please explore the Product Information Sheet (PIS) (/products/stemdiff-cerebral-organoid-kit.html#section-protocols-and-documentation) and Educational Materials (/products/stemdiff-cerebral-organoid-kit.html#section-resources-and-publications).
Catalog No. 50-197-4446 Supplier STEMCELL Technologies Supplier No. 08571
Culture pluripotent stem cell (PSC)-derived neural organoids for extended periods (> 40 days) with the STEMdiff™ Cerebral Organoid Maturation Kit.For your convenience, the maturation kit is fully compatible with the STEMdiff™ Cerebral Organoid Kit (/products/stemdiff-cerebral-organoid-kit.html), which can be used to generate neural organoids under defined and serum-free conditions.For more information on protocols for organoid culture with STEMdiff™ Cerebral Organoid Maturation Kit, please explore the Product Information Sheet (PIS) (/products/stemdiff-cerebral-organoid-kit.html#section-protocols-and-documentation) and Educational Materials (/products/stemdiff-cerebral-organoid-kit.html#section-resources-and-publications).