Learn More
STEMCELL Technologies STEMdiff™ Definitive Endoderm Kit, Size: 1 Kit
SDP
Catalog No. NC0626756
Encompass
Defined animal component-free medium for the differentiation of human ES and iPS cells to definitive endoderm
This item is not returnable. View return policy
Catalog No. NC0626756 Supplier STEMCELL Technologies Supplier No. 05110
Defined animal component-free medium for the differentiation of human ES and iPS cells to definitive endoderm