Learn More
STEMCELL Technologies STEMdiff™ Forebrain Neuron Differentiation Kit, Size: 1 Kit
SDP
Catalog No. 501974447
Encompass
The STEMdiff™ Forebrain Neuron Differentiation Kit is used in conjunction with the STEMdiff™ Forebrain Neuron Maturation Kit (Catalog #08605) to generate a mixed population of forebrain-type (FOXG1-positive) neurons from neural progenitor cells derived from human pluripotent stem cells. This kit is optimized to work with STEMdiff™ SMADi Neural Induction Kit, which supplies the appropriate neural progenitor cells. Neurons derived using these products are versatile tools for modeling human neurological development and disease, drug screening, toxicity testing, and cell therapy validation.
Catalog No. 50-197-4447 Supplier STEMCELL Technologies Supplier No. 08600
The STEMdiff™ Forebrain Neuron Differentiation Kit is used in conjunction with the STEMdiff™ Forebrain Neuron Maturation Kit (Catalog #08605) to generate a mixed population of forebrain-type (FOXG1-positive) neurons from neural progenitor cells derived from human pluripotent stem cells. This kit is optimized to work with STEMdiff™ SMADi Neural Induction Kit, which supplies the appropriate neural progenitor cells. Neurons derived using these products are versatile tools for modeling human neurological development and disease, drug screening, toxicity testing, and cell therapy validation.