Learn More
STEMCELL Technologies STEMdiff™ Hematopoietic - EB Basal Medium, Size: 120 mL
SDP
Catalog No. NC2043816
Encompass
Use STEMdiff™ Hematopoietic - EB Basal Medium in combination with STEMdiff™ Hematopoietic - EB Supplement A (/products/stemdiff-hematopoietic-eb-supplement-a.html) and STEMdiff™ Hematopoietic - EB Supplement B (/products/stemdiff-hematopoietic-eb-supplement-b.html) to support the differention of human pluripotent stem cells (hPSCs) to embroyid bodies.Available either individually or as part of the STEMdiff™ NK Cell Kit (/products/stemdiff-nk-cell-kit.html) or STEMdiff™ T Cell Kit (/products/stemdiff-t-cell-kit.html), where it can be used to promote the differentiation of hPSCs to NK or T cells, respectively. For detailed information on the two differentiation protocols, please explore the STEMdiff™ NK Cell Kit Technical Manual (/media/files/manual/10000007537-Generation_of_NK_Cells_Using_STEMdiff_or_StemSpan_NK_Cell_Kits.pdf) or the STEMdiff™ T Cell Kit Technical Manual (/media/files/pis/10000007545-PIS_02.pdf).
Catalog No. NC2043816 Supplier STEMCELL Technologies Supplier No. 1000171
Use STEMdiff™ Hematopoietic - EB Basal Medium in combination with STEMdiff™ Hematopoietic - EB Supplement A (/products/stemdiff-hematopoietic-eb-supplement-a.html) and STEMdiff™ Hematopoietic - EB Supplement B (/products/stemdiff-hematopoietic-eb-supplement-b.html) to support the differention of human pluripotent stem cells (hPSCs) to embroyid bodies.Available either individually or as part of the STEMdiff™ NK Cell Kit (/products/stemdiff-nk-cell-kit.html) or STEMdiff™ T Cell Kit (/products/stemdiff-t-cell-kit.html), where it can be used to promote the differentiation of hPSCs to NK or T cells, respectively. For detailed information on the two differentiation protocols, please explore the STEMdiff™ NK Cell Kit Technical Manual (/media/files/manual/10000007537-Generation_of_NK_Cells_Using_STEMdiff_or_StemSpan_NK_Cell_Kits.pdf) or the STEMdiff™ T Cell Kit Technical Manual (/media/files/pis/10000007545-PIS_02.pdf).