Learn More
STEMCELL Technologies STEMdiff Hepatocyte Kit, Size: 1 Kit
SDP
Catalog No. 502390442
Encompass
Efficiently generate hepatocyte-like cells (HLCs) that express several key liver-specific markers including albumin and alpha1-antitrypsin. The STEMdiff Hepatocyte Kit provides a reliable source of HLCs for your experiments by reproducibly differentiating human pluripotent stem cells (hPSCs) into HLCs across a variety of hPSC lines. The serum-free formulation minimizes experimental variability by reducing undefined components and enables you to robustly differentiate HLC cultures. HLCs generated using STEMdiff Hepatocyte Kit are suitable for a variety of applications in liver research disease modeling and hepatotoxicity testing.
Catalog No. 50-239-0442 Supplier STEMCELL Technologies Supplier No. 1000520
Efficiently generate hepatocyte-like cells (HLCs) that express several key liver-specific markers including albumin and alpha1-antitrypsin. The STEMdiff Hepatocyte Kit provides a reliable source of HLCs for your experiments by reproducibly differentiating human pluripotent stem cells (hPSCs) into HLCs across a variety of hPSC lines. The serum-free formulation minimizes experimental variability by reducing undefined components and enables you to robustly differentiate HLC cultures. HLCs generated using STEMdiff Hepatocyte Kit are suitable for a variety of applications in liver research disease modeling and hepatotoxicity testing.