Learn More
STEMCELL Technologies STEMdiff™ Intestinal Organoid Growth Medium, Size: 1 Kit
SDP
Catalog No. NC1641943
Encompass
Serum-free medium kit for maintenance of hPSC-derived human intestinal organoids
Catalog No. NC1641943 Supplier STEMCELL Technologies Supplier No. 05145
Serum-free medium kit for maintenance of hPSC-derived human intestinal organoids