Learn More
STEMCELL Technologies STEMdiff™ Neural Crest Differentiation Kit, undefined: 1 Kit
SDP
Catalog No. 501974449
Encompass
STEMdiff™ Neural Crest Differentiation Kit creates a serum-free medium for differentiation of human embryonic stem (ES) cells and induced pluripotent stem (iPS) cells to neural crest cells. These neural crest cells, which are characterized by neural crest markers such as SOX10 and CD271, can be differentiated to several downstream derivatives including chondrocytes, osteoblasts, and peripheral neurons. This medium is compatible with human ES and iPS cells maintained in either mTeSR™1 (Catalog #85850), mTeSR™ Plus (Catalog #100-0276) or TeSR™-E8™ (Catalog #05990).
Catalog No. 50-197-4449 Supplier STEMCELL Technologies Supplier No. 08610
STEMdiff™ Neural Crest Differentiation Kit creates a serum-free medium for differentiation of human embryonic stem (ES) cells and induced pluripotent stem (iPS) cells to neural crest cells. These neural crest cells, which are characterized by neural crest markers such as SOX10 and CD271, can be differentiated to several downstream derivatives including chondrocytes, osteoblasts, and peripheral neurons. This medium is compatible with human ES and iPS cells maintained in either mTeSR™1 (Catalog #85850), mTeSR™ Plus (Catalog #100-0276) or TeSR™-E8™ (Catalog #05990).