Learn More
STEMCELL Technologies STEMdiff™ Neural Induction Medium, Size: 250 mL
SDP
Catalog No. NC1238362
Encompass
STEMdiff™ Neural Induction Medium is a defined, serum-free medium for the neural induction of human embryonic stem (ES) cells and induced pluripotent stem (iPS) cells. This medium enables highly efficient generation of neural progenitor cell using either embryoid body- or monolayer culture-based protocols.
Learn how to generate neural progenitor cells from human pluripotent stem cells (hPSCs) in our On-Demand Neural Induction Course (https://www.stemcell.com/forms/neural-induction-on-demand-training.html), and browse our Tech Tips on the Neural Induction of hPSCs using the Embryoid Body Method (https://www.stemcell.com/technical-resources/educational-materials/tech-tips/obtain-hpsc-derived-npcs-using-an-eb-based-protocol-with-the-stemdiff-neural-induction-system.html) or Monolayer Method (https://www.stemcell.com/technical-resources/educational-materials/tech-tips/neural-induction-for-human-pluripotent-stem-cells-hpscs-using-monolayer-culture.html).
Catalog No. NC1238362 Supplier STEMCELL Technologies Supplier No. 05835
STEMdiff™ Neural Induction Medium is a defined, serum-free medium for the neural induction of human embryonic stem (ES) cells and induced pluripotent stem (iPS) cells. This medium enables highly efficient generation of neural progenitor cell using either embryoid body- or monolayer culture-based protocols.
Learn how to generate neural progenitor cells from human pluripotent stem cells (hPSCs) in our On-Demand Neural Induction Course (https://www.stemcell.com/forms/neural-induction-on-demand-training.html), and browse our Tech Tips on the Neural Induction of hPSCs using the Embryoid Body Method (https://www.stemcell.com/technical-resources/educational-materials/tech-tips/obtain-hpsc-derived-npcs-using-an-eb-based-protocol-with-the-stemdiff-neural-induction-system.html) or Monolayer Method (https://www.stemcell.com/technical-resources/educational-materials/tech-tips/neural-induction-for-human-pluripotent-stem-cells-hpscs-using-monolayer-culture.html).