Learn More
STEMCELL Technologies STEMdiff™ SMADi Neural Induction Kit, Size: 1 Kit
SDP
Catalog No. NC1460118
Encompass
Serum-free medium kit for highly efficient SMAD inhibition-mediated neural induction of human ES and iPS cells
Catalog No. NC1460118 Supplier STEMCELL Technologies Supplier No. 08581
Serum-free medium kit for highly efficient SMAD inhibition-mediated neural induction of human ES and iPS cells