Learn More
STEMCELL Technologies STEMdiff™ Trilineage Differentiation Kit, Size: 1 Kit
SDP
Catalog No. NC1161572
Encompass
The STEMdiff™ Trilineage Differentiation Kit provides a simple culture assay to functionally validate the ability of new or established human embryonic stem (ES) and induced pluripotent stem (iPS) cell lines to differentiate to the three germ layers: ectoderm, mesoderm, and endoderm. This kit includes specialized, complete media and monolayer-based protocols to perform parallel in vitro directed differentiation experiments for each germ layer, clearly and reproducibly establishing trilineage differentiation potential within one week. STEMdiff™ Trilineage Differentiation Kit is intended to be an endpoint assay and is not optimized for the generation of cells for downstream differentiation or other applications. STEMdiff™ Trilineage Differentiation Kit has been optimized to assess cells maintained in mTeSR™1.
Catalog No. NC1161572 Supplier STEMCELL Technologies Supplier No. 05230
Add to Cart
Add to Cart
The STEMdiff™ Trilineage Differentiation Kit provides a simple culture assay to functionally validate the ability of new or established human embryonic stem (ES) and induced pluripotent stem (iPS) cell lines to differentiate to the three germ layers: ectoderm, mesoderm, and endoderm. This kit includes specialized, complete media and monolayer-based protocols to perform parallel in vitro directed differentiation experiments for each germ layer, clearly and reproducibly establishing trilineage differentiation potential within one week. STEMdiff™ Trilineage Differentiation Kit is intended to be an endpoint assay and is not optimized for the generation of cells for downstream differentiation or other applications. STEMdiff™ Trilineage Differentiation Kit has been optimized to assess cells maintained in mTeSR™1.