Learn More
Amsbio LLC StemFit Basic04 Complete (AOF, defined medium for human pluripotent stem cell culture, including 80ng/ml bFGF)
Catalog No. 502041689
Encompass
StemFit is a range of animal-origin free, chemically defined media proven to effectively maintain induced pluripotent stem cells (iPSCs) and embryonic stem cells (ESCs) under feeder-free conditions during the reprogramming, expansion and differentiation phases of stem cell culture and growth medium. StemFit Basic04 MAIN FEATURES: Animal origin-free medium for the maintenance and expansion of human pluripotent stem cells | Contains 80 ng/mL basic FGF | Simplified one bottle formula increases guarantee of sterilization | Suitable for both basic and clinical research | Superior expansion with single-cell passaging | Chemically defined formula minimizes lot-to-lot variation | Consistent high performance with various matrices | Weekend-free cell culture | Reduced media consumption | Reproducible growth rates.
Catalog No. 50-204-1689 Supplier Amsbio LLC Supplier No. SF041001
StemFit is a range of animal-origin free, chemically defined media proven to effectively maintain induced pluripotent stem cells (iPSCs) and embryonic stem cells (ESCs) under feeder-free conditions during the reprogramming, expansion and differentiation phases of stem cell culture and growth medium. StemFit Basic04 MAIN FEATURES: Animal origin-free medium for the maintenance and expansion of human pluripotent stem cells | Contains 80 ng/mL basic FGF | Simplified one bottle formula increases guarantee of sterilization | Suitable for both basic and clinical research | Superior expansion with single-cell passaging | Chemically defined formula minimizes lot-to-lot variation | Consistent high performance with various matrices | Weekend-free cell culture | Reduced media consumption | Reproducible growth rates.