Learn More
Amsbio LLC StemFit For Differentiation (AOF, chemically defined medium for human pluripotent stem cell culture)
Catalog No. 502563589
Encompass
StemFit For Differentiation (“StemFit Diff”) is a chemically defined, human/animal component-free supplement for differentiating human embryonic stem (ES) and induced pluripotent stem (iPS) cells into multiple lineages. It supports differentiation along ectoderm, mesoderm, or endoderm lineages with various induction factors or cytokines. StemFit Diff can be combined with StemFit Basic media and iMatrix-511 laminin as an ECM for clinical research applications of human pluripotent stem cell (hPSC)-derived cells/tissues, providing defined and xeno-free culture systems for expansion and differentiation. StemFit Diff is a 5X concentrate for use with basal medium (DMEM, DMEM/F12, RPMI1640, etc.) and induction factors or cytokines. MAIN FEATURES | Differentiates human ES and iPS cells to multiple lineages | Chemically defined to minimize lot-to-lot variation and ensure consistent cell differentiation | AOF composition minimizes risk of immunogenic contamination.
Catalog No. 50-256-3589 Supplier Amsbio LLC Supplier No. SFD401
Add to Cart
Add to Cart
StemFit For Differentiation (“StemFit Diff”) is a chemically defined, human/animal component-free supplement for differentiating human embryonic stem (ES) and induced pluripotent stem (iPS) cells into multiple lineages. It supports differentiation along ectoderm, mesoderm, or endoderm lineages with various induction factors or cytokines. StemFit Diff can be combined with StemFit Basic media and iMatrix-511 laminin as an ECM for clinical research applications of human pluripotent stem cell (hPSC)-derived cells/tissues, providing defined and xeno-free culture systems for expansion and differentiation. StemFit Diff is a 5X concentrate for use with basal medium (DMEM, DMEM/F12, RPMI1640, etc.) and induction factors or cytokines. MAIN FEATURES | Differentiates human ES and iPS cells to multiple lineages | Chemically defined to minimize lot-to-lot variation and ensure consistent cell differentiation | AOF composition minimizes risk of immunogenic contamination.